Invitrogen
Manufacturer Code:PA5102098
Catalog # PIPA5102098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HRH1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bordetella pertussis induced histamine sensitization Bphs H1 receptor H1R H1-R HH1R Hir hisH1 Hisr Histamine H1 receptor Histamine H1-R histamine receptor histamine receptor H 1 histamine receptor H1 histamine receptor subclass H1 Hrh1; H1R; Histamine H1 receptor; histamine receptor, subclass H1
Gene Aliases: H1-R; H1R; HH1R; hisH1; HRH1
UniProt ID: (Human) P35367
Entrez Gene ID: (Human) 3269
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.