Proteintech
Manufacturer Code:134131AP150UL
Catalog # 50-172-8725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HRH1 Fusion Protein Ag4067 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bordetella pertussis induced histamine sensitization; H1 R H1R HH1R hisH1 Histamine H1 receptor histamine receptor H1 HRH1; H1R; HH1R; Histamine H1 receptor; histamine receptor, subclass H1
Gene Aliases: Bphs; H1-R; H1R; HH1R; Hir; hisH1; Hisr; HRH1
UniProt ID: (Human) P35367, (Mouse) P70174, (Rat) P31390
Entrez Gene ID: (Human) 3269, (Mouse) 15465, (Rat) 24448
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.