Invitrogen
Manufacturer Code:PA597051
Catalog # PIPA597051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 20-260 of human HRG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; Histidine-rich glycoprotein 1; HPRG; HRG1
Gene Aliases: AI265597; AW413091; D16jh2; D18020; HPRG; Hrg; HRG1; Hrgp
UniProt ID: (Rat) Q99PS8, (Mouse) Q9ESB3
Entrez Gene ID: (Rat) 171016, (Mouse) 94175
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.