Invitrogen
Manufacturer Code:PA5110899
Catalog # PIPA5110899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human HRD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200010C09Rik AW211966 C85322 D530017H19Rik DER3 E3 ubiquitin-protein ligase synoviolin HRD1 HRD1 Protein KIAA1810 MGC40372 RGD1310488 RING-type E3 ubiquitin transferase synoviolin Synovial apoptosis inhibitor 1 Syvn1; E3 ubiquitin-protein ligase synoviolin; HMG-coA reductase degradation 1 homolog; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin
Gene Aliases: DER3; HRD1; KIAA1810; SYVN1
UniProt ID: (Human) Q86TM6
Entrez Gene ID: (Human) 84447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.