Invitrogen
Manufacturer Code:PA5100081
Catalog # PIPA5100081
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SYVN1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200010C09Rik AW211966 C85322 D530017H19Rik DER3 E3 ubiquitin-protein ligase synoviolin HMG-coA reductase degradation 1 homolog HRD1 HRD1 protein KIAA1810 MGC40372 RGD1310488 RING-type E3 ubiquitin transferase synoviolin Synovial apoptosis inhibitor 1 synovial apoptosis inhibitor 1 synoviolin synoviolin 1 SYVN1; E3 ubiquitin-protein ligase synoviolin; HMG-coA reductase degradation 1 homolog; HRD1 protein; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin; synoviolin 1
Gene Aliases: 1200010C09Rik; AW211966; C85322; D530017H19Rik; DER3; HRD1; KIAA1810; RGD1310488; SYVN1
UniProt ID: (Human) Q86TM6, (Mouse) Q9DBY1
Entrez Gene ID: (Human) 84447, (Mouse) 74126, (Rat) 361712
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.