Novus Biologicals
Manufacturer Code:NBP302998100UL
Catalog # NB093560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human HRD1 (NP_757385.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase synoviolin; E3 ubiquitin-protein ligase synoviolin EC 6.3.2 HRD1EC 6.3.2.- KIAA1810Synovial apoptosis inhibitor 1 MGC40372 synovial apoptosis inhibitor 1 synoviolin; HMG-coA reductase degradation 1 homolog; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin
Gene Aliases: DER3; HRD1; KIAA1810; SYVN1
UniProt ID: (Human) Q86TM6
Entrez Gene ID: (Human) 84447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.