Proteintech
Manufacturer Code:175811AP150UL
Catalog # 50-173-1664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HRASLS5 Fusion Protein Ag11113 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ca(2+)-independent N-acyltransferase; Ca-independent N-acyltransferase; calcium-independent phosphatidylethanolamine N-acyltransferase; H rev107 like protein 5 HRAS like suppressor 5 HRASLS5 HRLP5 iNAT RLP1; H-rev107-like protein 5; HRAS-like suppressor 5; HRAS-like suppressor family, member 5; HRSL5; iNAT; lecithin-retinol acyltransferase (LRAT)-like protein-1; Phospholipase A and acyltransferase 5; Rat LRAT-like protein-1; RLP-1; testicular tissue protein Li 90
Gene Aliases: 1700055A14Rik; 1700086E08Rik; 4921526K24Rik; HRASLS5; HRLP5; HRSL5; iNAT; PLAAT5; RGD1308376; Rlp-1; RLP1
UniProt ID: (Human) Q96KN8, (Rat) Q4KLN5, (Mouse) Q9CPX5
Entrez Gene ID: (Human) 117245, (Rat) 293711, (Mouse) 66727
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.