Abnova
Manufacturer Code:H00117245K
Catalog # 89-207-915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human HRASLS5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human HRASLS5 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Ca(2+)-independent N-acyltransferase; calcium-independent phosphatidylethanolamine N-acyltransferase; H-rev107-like protein 5; HRAS-like suppressor 5; HRAS-like suppressor family, member 5; HRLP5 RLP-1 RLP1; HRSL5; iNAT; lecithin-retinol acyltransferase (LRAT)-like protein-1; Phospholipase A and acyltransferase 5; testicular tissue protein Li 90
Gene Aliases: HRASLS5; HRLP5; HRSL5; iNAT; PLAAT5; RLP1
UniProt ID: (Human) Q96KN8
Entrez Gene ID: (Human) 117245
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.