Novus Biologicals
Manufacturer Code:NBP271337V
Catalog # NBP271337V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E11 |
Immunogen | Full length recombinant protein of human HR6B/UBE2B (NP_003328) produced in E.coli. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: E2 protein; E2 protein E2-17kDa HHR6B HR6B RAD6 homolog B RAD6BEC 6.3.2.19 UBC2 Ubiquitin carrier protein B ubiquitin-conjugating enzyme E2 B Ubiquitin-conjugating enzyme E2-17 kDa ubiquitin-conjugating enzyme E2B (RAD6 homolog) Ubiquitin-protein ligase B; E2 ubiquitin-conjugating enzyme B; HR6B; RAD6 homolog B; Ubiquitin carrier protein B; ubiquitin conjugating enzyme E2B; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B (RAD6 homolog); Ubiquitin-protein ligase B
Gene Aliases: E2-17kDa; HHR6B; HR6B; RAD6B; UBC2; UBE2B
UniProt ID: (Human) P63146
Entrez Gene ID: (Human) 7320
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.