Invitrogen
Manufacturer Code:PA517592
Catalog # PIPA517592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids at the amino-terminus of human HR6A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG2013-PA; CG2013-PC; E2 protein; E2 ubiquitin-conjugating enzyme 6; E2 ubiquitin-conjugating enzyme A; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; HR6A; HR6B; RAD6 homolog A; RAD6 homolog B; RE56673p; Ubc6-PA; Ubc6-PC; Ube2a ube2b Ubiquitin-conjugating enzyme E2 A RAD6 homolog A hHR6A Ubiquitin carrier protein A Ubiquitin-protein ligase A RAD6A Ubiquitin-conjugating enzyme E2 B RAD6 homolog B hHR6B Ubiquitin carrier protein B Ubiquitin-conjugating enzyme E2-17 kDa Ubiquitin-protein ligase B RAD6B; Ubiquitin carrier protein; Ubiquitin carrier protein A; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2A; ubiquitin conjugating enzyme E2B; ubiquitin conjugating enzyme E2B S homeolog; Ubiquitin-conjugating enzyme E2 A; Ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); Ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2A, RAD6 homolog; ubiquitin-conjugating enzyme E2B; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-conjugating enzyme HR6A; Ubiquitin-protein ligase; Ubiquitin-protein ligase A; Ubiquitin-protein ligase B
Gene Aliases: 2610301N02Rik; BcDNA:RE56673; CG2013; Dhr6; Dmel\CG2013; Dmel_CG2013; dRAD6; E2-14k; E2-17kDa; HHR6A; HHR6B; HR6A; HR6B; Mhr6a; mHR6B; mp:zf637-2-000771; MRXS30; MRXSN; RAD6; RAD6A; RAD6B; si:bz46j2.4; UBC2; UBC6; UbcD6; UBE2A; UBE2B; wu:fa01h11; XELAEV_18019992mg; XELAEV_18038882mg; zgc:100921
UniProt ID: (Human) P49459, (Human) P63146, (Fruit fly) P25153, (Mouse) Q9Z255, (Mouse) P63147, (Rat) P63149
Entrez Gene ID: (Human) 7319, (Chicken) 395672, (Human) 7320, (Chicken) 100857678, (Fruit fly) 40610, (Rat) 298317, (Mouse) 22209, (Mouse) 22210, (Rat) 81816, (Zebrafish) 797853, (Xenopus) 398788, (Zebrafish) 437020, (Xenopus) 443745
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.