Proteintech
Manufacturer Code:166731AP150UL
Catalog # 50-173-1103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HPSE Fusion Protein Ag10167 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endo glucoronidase HEP heparanase Heparanase 1 HPA Hpa1 HPR1 HPSE HPSE1 HSE1; Endo-glucoronidase; endoglycosidase heparanase; Heparanase; Heparanase 50 kDa subunit; Heparanase 8 kDa subunit; Heparanase-1; heparanase-like; Hpa1
Gene Aliases: HEP; HPA; HPA1; HPR1; HPSE; HPSE1; HSE1
UniProt ID: (Human) Q9Y251, (Mouse) Q6YGZ1, (Rat) Q71RP1
Entrez Gene ID: (Human) 10855, (Mouse) 15442, (Rat) 64537
Molecular Function:
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.