Invitrogen
Manufacturer Code:PA595699
Catalog # PIPA595699
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 550-650 of human HPS4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hermansky-Pudlak syndrome 4; Hermansky-Pudlak syndrome 4 homolog; Hermansky-Pudlak syndrome 4 protein; Hermansky-Pudlak syndrome 4 protein homolog; Le protein; light ear; Light-ear protein; Light-ear protein homolog
Gene Aliases: 2010205O06Rik; AU040608; BLOC-3; BLOC3S2; C130020P05Rik; HPS4; KIAA1667; LE; mKIAA1667
UniProt ID: (Human) Q9BY93, (Mouse) Q99KG7
Entrez Gene ID: (Human) 89781, (Rat) 304555, (Mouse) 192232
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.