Novus Biologicals
Manufacturer Code:NBP231898
Catalog # NBP231898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: GQRGNKLPTGEQGLDEDVDGVCESHAAPGLECSSGSANCQGAGPSADGISSRLTPAESCMGLVRMNLYTHCVKGLVLSLLAEEPLLGDSAAIEEVYHSSLASLNGLEVHLKETLPRDEAASTSSTYNFTHYDR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bK1048E9.4 Hermansky-Pudlak syndrome 4 Hermansky-Pudlak syndrome 4 protein KIAA1667bK1048E9.5 LE Light-ear protein homolog; Hermansky-Pudlak syndrome 4; Hermansky-Pudlak syndrome 4 protein; Light-ear protein homolog
Gene Aliases: BLOC3S2; HPS4; KIAA1667; LE
UniProt ID: (Human) Q9BY93
Entrez Gene ID: (Human) 89781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.