Novus Biologicals
Manufacturer Code:NBP212359C
Catalog # NBP212359C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0331-6Z48 |
Immunogen | Human HRG |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp779H1622 Histidine-proline-rich glycoprotein histidine-rich glycoprotein HPRGHRGP; Histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; HPRG
Gene Aliases: HPRG; HRG; HRGP; THPH11
UniProt ID: (Human) P04196
Entrez Gene ID: (Human) 3273
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.