R&D Systems
Manufacturer Code:MAB1869SP
Catalog # MAB1869SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 227901 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human HPRG Val19-Lys525 Accession # P04196 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp779H1622 Histidine-proline-rich glycoprotein histidine-rich glycoprotein HPRGHRGP HRG; Histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; HPRG
Gene Aliases: HPRG; HRG; HRGP; THPH11
UniProt ID: (Human) P04196
Entrez Gene ID: (Human) 3273
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.