Invitrogen
Manufacturer Code:PA5106522
Catalog # PIPA5106522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MEKKK 1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11.1 Hematopoietic progenitor kinase hematopoietic progenitor kinase 1 HPK HPK1 Map4k1 MAPK/ERK kinase kinase kinase 1 MEK kinase kinase 1 MEKKK 1 mHPK1 mitogen activated protein kinase kinase kinase kinase 1 mitogen-activated protein kinase kinase kinase kinase 1; Hematopoietic progenitor kinase; hematopoietic progenitor kinase 1; HPK; MAPK/ERK kinase kinase kinase 1; MEK kinase kinase 1; MEKKK 1; mitogen activated protein kinase kinase kinase kinase 1; Mitogen-activated protein kinase kinase kinase kinase 1
Gene Aliases: HPK1; MAP4K1; mHPK1
UniProt ID: (Human) Q92918, (Mouse) P70218
Entrez Gene ID: (Human) 11184, (Mouse) 26411
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.