Invitrogen
Manufacturer Code:PA554958
Catalog # PIPA554958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HPGDS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glutathione S-transferase; glutathione S-transferase sigma; Glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; Glutathione-requiring prostaglandin D synthase; GST class-sigma; GSTS PGDS PTGDS2 EC 2.5.1.18 EC 5.3.99.2 Glutathione S-transferase glutathione S-transferase sigma Glutathione-dependent PGD synthase Glutathione-requiring prostaglandin D synthase GST class-sigma GSTShematopoietic prostaglandin D2 synthase hematopoietic prostaglandin D synthase H-PGDS PGDSglutathione-dependent PGD synthetase Prostaglandin-H2 D-isomerase PTGDS2; H-PGDS; Hematopoietic prostaglandin D synthase; hematopoietic prostaglandin D2 synthase; Prostaglandin-H2 D-isomerase
Gene Aliases: GSTS; GSTS1; GSTS1-1; HPGDS; PGD2; PGDS; PTGDS2
UniProt ID: (Human) O60760
Entrez Gene ID: (Human) 27306
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.