Invitrogen
Manufacturer Code:PA543217
Catalog # PIPA543217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PGDS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glutathione S-transferase; glutathione S-transferase sigma; Glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; Glutathione-requiring prostaglandin D synthase; GST class-sigma; H-PGDS; Hematopoietic prostaglandin D synthase; Hematopoietic prostaglandin D synthase GSTS PGD2 PGDS GSTS1-1; hematopoietic prostaglandin D2 synthase; prostaglandin D2 synthase 2, hematopoietic; prostaglandin D2 synthase, hematopoietic; Prostaglandin-H2 D-isomerase
Gene Aliases: GSTS; GSTS1; GSTS1-1; H-PGDS; HPGDS; PGD2; PGDS; PTGDS2
UniProt ID: (Human) O60760, (Rat) O35543, (Mouse) Q9JHF7
Entrez Gene ID: (Horse) 100053460, (Human) 27306, (Dog) 478481, (Rabbit) 100353701, (Rat) 58962, (Mouse) 54486, (Guinea pig) 100734640
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.