Proteintech
Manufacturer Code:225221AP150UL
Catalog # 50-173-3880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HPGDS Fusion Protein Ag18290 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glutathione S transferase GST class sigma GSTS H PGDS HPGDS PGDS Prostaglandin H2 D isomerase PTGDS2; Glutathione S-transferase; glutathione S-transferase sigma; Glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; Glutathione-requiring prostaglandin D synthase; GST class-sigma; H-PGDS; Hematopoietic prostaglandin D synthase; hematopoietic prostaglandin D2 synthase; prostaglandin D2 synthase 2, hematopoietic; Prostaglandin-H2 D-isomerase
Gene Aliases: GSTS; GSTS1; GSTS1-1; H-PGDS; HPGDS; PGD2; PGDS; PTGDS2
UniProt ID: (Human) O60760, (Mouse) Q9JHF7
Entrez Gene ID: (Human) 27306, (Mouse) 54486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.