OriGene
Manufacturer Code:TA700099
Catalog # 50-167-7636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C10 |
Immunogen | Full length human recombit protein of human HPGD produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; 15-PGDH PGDH PGDH1 PHOAR1 SDR36C1; NAD+-dependent 15-hydroxyprostaglandin dehydrogenase; Prostaglandin dehydrogenase 1; Short chain dehydrogenase/reductase family 36C member 1; short chain dehydrogenase/reductase family 36C, member 1
Gene Aliases: 15-PGDH; HPGD; PGDH; PGDH1; PHOAR1; SDR36C1
UniProt ID: (Human) P15428
Entrez Gene ID: (Human) 3248
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.