Invitrogen
Manufacturer Code:PA556898
Catalog # PIPA556898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HPDL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-HPPD-L GLOXD1 RP4-534D1.1 4-hydroxyphenylpyruvate dioxygenase-like protein glyoxalase domain containing 1 glyoxalase domain-containing protein 1; 4-hydroxyphenylpyruvate dioxygenase-like protein; glyoxalase domain containing 1; Glyoxalase domain-containing protein 1; HPD-like protein
Gene Aliases: 4-HPPD-L; GLOXD1; HPDL
UniProt ID: (Human) Q96IR7
Entrez Gene ID: (Human) 84842
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.