Proteintech
Manufacturer Code:207771AP150UL
Catalog # 50-173-3077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HPDL Fusion Protein Ag14773 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4 HPPD L GLOXD1 HPDL RP4 534D1.1; 4-hydroxyphenylpyruvate dioxygenase-like protein; glyoxalase domain containing 1; Glyoxalase domain-containing protein 1; HPD-like protein
Gene Aliases: 4-HPPD-L; GLOXD1; HPDL
UniProt ID: (Human) Q96IR7
Entrez Gene ID: (Human) 84842
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.