Invitrogen
Manufacturer Code:PA558343
Catalog # PIPA558343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HPD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-HPPD 4HPPD GLOD3 HPPDASE PPD 4-hydroxyphenylpyruvic acid oxidase glyoxalase domain containing 3; 4-hydroxyphenylpyruvate dioxygenase; 4-hydroxyphenylpyruvic acid dioxygenase; 4-hydroxyphenylpyruvic acid oxidase; 4HPPD; F Alloantigen; F liver antigen; F protein; glyoxalase domain containing 3; HPPDase
Gene Aliases: 4-HPPD; 4HPPD; Fla; Flp; GLOD3; HPD; Hppd; HPPDASE; Laf; PPD
UniProt ID: (Human) P32754, (Mouse) P49429, (Rat) P32755
Entrez Gene ID: (Human) 3242, (Mouse) 15445, (Rat) 29531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.