Novus Biologicals
Manufacturer Code:NBP299592AF594
Catalog # NB061938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HPD. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-hydroxyphenylpyruvate dioxygenase; 4-hydroxyphenylpyruvic acid oxidase; 4HPPD; 4HPPDEC 1.13.11.274-HPPD 4-hydroxyphenylpyruvate dioxygenase GLOD3 glyoxalase domain containing 34-hydroxyphenylpyruvic acid oxidase HPPDase PPD; glyoxalase domain containing 3
Gene Aliases: 4-HPPD; 4HPPD; GLOD3; HPD; HPPDASE; PPD
UniProt ID: (Human) P32754
Entrez Gene ID: (Human) 3242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.