Invitrogen
Manufacturer Code:PA521326
Catalog # PIPA521326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fragment corresponding to a region within amino acids 260 and 582 of HPA2 (Uniprot ID#Q8WWQ2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles,Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: heparanase 3; heparanase-like protein; Hpa2; HPA2 HPR2 UFS heparanase 3 heparanase-2 heparanase-like protein; Inactive heparanase-2
Gene Aliases: Gm968; HPA2; HPR2; HPSE2; UFS; UFS1
UniProt ID: (Human) Q8WWQ2, (Mouse) B2RY83
Entrez Gene ID: (Human) 60495, (Mouse) 545291
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.