Novus Biologicals
Manufacturer Code:NBP155329
Catalog # NBP155329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to HP1BP3(heterochromatin protein 1 binding protein 3) The peptide sequence was selected from the N terminal of HP1BP3. Peptide sequence SEESVSTVEEQENETPPATSSEAEQPKGEPENEEKEENKSSEETKKDEKD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: heterochromatin protein 1 binding protein 3 heterochromatin protein 1-binding protein 3 HP1-BP74 MGC43701 Protein HP1-BP74; heterochromatin protein 1, binding protein 3; Heterochromatin protein 1-binding protein 3; HP1-BP74; Protein HP1-BP74
Gene Aliases: HP1-BP74; HP1BP3; HP1BP74
UniProt ID: (Human) Q5SSJ5
Entrez Gene ID: (Human) 50809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.