Invitrogen
Manufacturer Code:PA568811
Catalog # PIPA568811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse HOXD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box D3; homeo box D4; Homeobox D3 HOX1D HOX4 HOX4A Hox-4.1 MGC10470; Homeobox gene D3; Homeobox protein Hox-4.1; Homeobox protein Hox-4A; Homeobox protein Hox-D3; Homeobox protein MH-19; homeobox protein R6; Hox-4.1, mouse, homolog of
Gene Aliases: Hox-4.1; Hox-5.5; HOX1D; HOX4; HOX4A; Hox4r6; Hoxd-3; HOXD3
UniProt ID: (Human) P31249, (Mouse) P09027
Entrez Gene ID: (Horse) 100053542, (Dog) 488417, (Human) 3232, (Rabbit) 100337724, (Rat) 288152, (Guinea pig) 100716121, (Mouse) 15434
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.