Invitrogen
Manufacturer Code:PA567561
Catalog # PIPA567561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human HOXD3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box D3; homeo box D4; Homeobox D3 HOX1D HOX4 HOX4A Hox-4.1 MGC10470; Homeobox protein Hox-4.1; Homeobox protein Hox-4A; Homeobox protein Hox-D3; Homeobox protein MH-19; Hox-4.1, mouse, homolog of
Gene Aliases: Hox-4.1; Hox-5.5; HOX1D; HOX4; HOX4A; Hoxd-3; HOXD3
UniProt ID: (Human) P31249, (Mouse) P09027
Entrez Gene ID: (Human) 3232, (Mouse) 15434
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.