Novus Biologicals
Manufacturer Code:NBP174094
Catalog # NBP174094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the middle region of HOXD3. Immunizing peptide sequence AHKGAELNGSCMRPGTGNSQGGGGGSQPPGLNSEQQPPQPPPPPPTLPPS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box D3; homeo box D3 homeobox D3 Hox-4.1 HOX4AMGC10470 mouse homolog of; Homeobox protein Hox-4A; Homeobox protein Hox-D3; Hox-4.1, mouse, homolog of
Gene Aliases: Hox-4.1; HOX1D; HOX4; HOX4A; HOXD3
UniProt ID: (Human) P31249
Entrez Gene ID: (Human) 3232
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.