Invitrogen
Manufacturer Code:PA568541
Catalog # PIPA568541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse HOXD13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BDE BDSD HOX4I SPD homeo box 4I homeo box D13 homeobox protein Hox-4I homeobox protein Hox-D13; homeo box 4I; homeo box D13; Homeobox protein Hox-4.8; Homeobox protein Hox-4I; Homeobox protein Hox-D13
Gene Aliases: BDE; BDSD; Hox-4.8; HOX4I; HOXD13; SPD; SPD1; spdh
UniProt ID: (Human) P35453, (Mouse) P70217
Entrez Gene ID: (Rabbit) 100301549, (Human) 3239, (Goat) 100860842, (Horse) 100146425, (Rat) 288154, (Mouse) 15433
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.