Novus Biologicals
Manufacturer Code:NBP152149
Catalog # NBP152149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KSSFPGDVALNQPD corresponding to internal region according to NP_000514.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BDSD homeo box 4I homeo box D13 homeobox D13 Homeobox protein Hox-4I homeobox protein Hox-D13 HOX4IBDE SPD; homeo box 4I; homeo box D13; Homeobox protein Hox-4I; Homeobox protein Hox-D13
Gene Aliases: BDE; BDSD; HOX4I; HOXD13; SPD; SPD1
UniProt ID: (Human) P35453
Entrez Gene ID: (Human) 3239
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.