Proteintech
Manufacturer Code:187341AP150UL
Catalog # 50-173-2217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 4F; homeo box D11; homeobox D11 Homeobox protein Hox 4F Homeobox protein Hox D11 HOX4 HOX4F HOXD11; Homeobox protein Hox-4.6; Homeobox protein Hox-4F; Homeobox protein Hox-5.5; Homeobox protein Hox-D11; Hox-4.6, mouse, homolog of; uncharacterized LOC102553842
Gene Aliases: E230017H14Rik; Hox-4.6; Hox-5.4; Hox-5.5; HOX4; HOX4F; Hoxd-11; HOXD11
UniProt ID: (Human) P31277, (Mouse) P23813
Entrez Gene ID: (Human) 3237, (Mouse) 15431, (Rat) 102553842
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.