Novus Biologicals
Manufacturer Code:NBP270920
Catalog # NBP270920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D11 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 80-340 of human HOXD10 (NP_002139) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: homeo box 4D; homeo box 4D homeo box D10 homeobox D10 Homeobox protein Hox-4D Homeobox protein Hox-4E homeobox protein Hox-D10 Hox-4.4 HOX4DHOX4 HOX4E; homeo box D10; Homeobox protein Hox-4D; Homeobox protein Hox-4E; Homeobox protein Hox-D10
Gene Aliases: Hox-4.4; HOX4; HOX4D; HOX4E; HOXD10
UniProt ID: (Human) P28358
Entrez Gene ID: (Human) 3236
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.