OriGene
Manufacturer Code:CF809671
Catalog # 50-166-9645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A6 |
Immunogen | Full length human recombit protein of human HOXC4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: homeo box 3E; Homeobox protein CP19; Homeobox protein Hox-3E; Homeobox protein Hox-C4; HOX3 HOX3E cp19
Gene Aliases: cp19; HOX3; HOX3E; HOXC4
UniProt ID: (Human) P09017
Entrez Gene ID: (Human) 3221
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.