Proteintech
Manufacturer Code:143211AP150UL
Catalog # 50-172-9363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HOXC4 Fusion Protein Ag5563 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cp19 homeobox C4 Homeobox protein CP19 Homeobox protein Hox 3E Homeobox protein Hox C4 HOX3 HOX3E HOXC4; homeo box 3E; homeo box C4; Homeobox gene C4; Homeobox protein CP19; Homeobox protein Hox-3.5; Homeobox protein Hox-3E; Homeobox protein Hox-C4; homeobox protein R3
Gene Aliases: cp19; Hox-3.5; HOX3; HOX3E; Hox3r3; Hoxc-4; HOXC4
UniProt ID: (Human) P09017, (Mouse) Q08624
Entrez Gene ID: (Human) 3221, (Mouse) 15423, (Rat) 24459
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.