Novus Biologicals
Manufacturer Code:NBP246000
Catalog # NBP246000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C10 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 1-262 of human HOXC10(NP_059105) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: homeo box C10; homeo box C10 homeobox C10 Homeobox protein Hox-3I homeobox protein Hox-C10 homeoprotein C10 HOX3I MGC5259; Homeobox protein Hox-3I; Homeobox protein Hox-C10; homeoprotein C10
Gene Aliases: HOX3I; HOXC10
UniProt ID: (Human) Q9NYD6
Entrez Gene ID: (Human) 3226
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.