Invitrogen
Manufacturer Code:PA568702
Catalog # PIPA568702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human HOXB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 2H; homeo box B2; homeo box B2-like; homeobox B2; Homeobox protein Hox-2.8; Homeobox protein Hox-2H; Homeobox protein Hox-B2; Homeobox protein Hox-B2 Homeobox protein Hox-2.8 Homeobox protein Hox-2H K8 HOX2H HOXB2; Hoxb2-like epididymal protein; K8; K8 home protein
Gene Aliases: AI894218; Hox-2.8; HOX2; HOX2H; HOXB2; Hoxbes2; K8
UniProt ID: (Human) P14652, (Mouse) P0C1T1
Entrez Gene ID: (Rabbit) 100347179, (Dog) 608880, (Horse) 100146636, (Bovine) 616129, (Human) 3212, (Guinea pig) 100720228, (Mouse) 103889, (Rat) 100361765
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.