Novus Biologicals
Manufacturer Code:NBP232338
Catalog # NBP232338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This HOXB2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 282-306 amino acids from the C-terminal region of human HOXB2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 2H; homeo box 2H homeo box B2 homeobox B2 Homeobox protein Hox-2.8 Homeobox protein Hox-2H homeobox protein Hox-B2 HOX2 HOX2HHox-2.8 K8 K8 home protein; homeo box B2; Homeobox protein Hox-2.8; Homeobox protein Hox-2H; Homeobox protein Hox-B2; K8; K8 home protein
Gene Aliases: Hox-2.8; HOX2; HOX2H; HOXB2; K8
UniProt ID: (Human) P14652
Entrez Gene ID: (Human) 3212
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.