Invitrogen
Manufacturer Code:PA572217
Catalog # PIPA572217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 182-315 amino acids from the C-terminal region of Mouse Hoxb1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box B1; Homeobox protein Hox-2.9; Homeobox protein Hox-2I; Homeobox protein Hox-B1; Homeobox protein Hox-B1 Homeobox protein Hox-29 Hoxb1 Hox-29 Hoxb-1
Gene Aliases: HCFP3; Hox-2.9; HOX2; HOX2I; Hoxb-1; HOXB1
UniProt ID: (Human) P14653, (Mouse) P17919
Entrez Gene ID: (Human) 3211, (Mouse) 15407
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.