R&D Systems
Manufacturer Code:AF6318SP
Catalog # AF6318SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human HOXB1 Met1-Thr192 Accession # P14653 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 2I homeo box B1 homeobox B1 Homeobox protein Hox-2I homeobox protein Hox-B1 HOX2 Hox-2.9 HOX2I HOX2IMGC116844 MGC116843 MGC116845; Homeobox protein Hox-2I; Homeobox protein Hox-B1
Gene Aliases: HCFP3; Hox-2.9; HOX2; HOX2I; HOXB1
UniProt ID: (Human) P14653
Entrez Gene ID: (Human) 3211
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.