Proteintech
Manufacturer Code:187321AP150UL
Catalog # 50-173-2216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box B1; homeobox B1 Homeobox protein Hox 2I Homeobox protein Hox B1 Hox 2.9 HOX2 HOX2I HOXB1; Homeobox protein Hox-2.9; Homeobox protein Hox-2I; Homeobox protein Hox-B1
Gene Aliases: HCFP3; Hox-2.9; HOX2; HOX2I; Hoxb-1; HOXB1
UniProt ID: (Human) P14653, (Mouse) P17919
Entrez Gene ID: (Human) 3211, (Mouse) 15407
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.