Invitrogen
Manufacturer Code:PA569008
Catalog # PIPA569008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human HOXA5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 1C; homeo box 1C homeo box A5 homeobox A5 Homeobox protein Hox-1C Homeobox protein Hox-A5; homeo box A5; Homeobox protein Hox-1.3; Homeobox protein Hox-1C; Homeobox protein Hox-A5; homeobox protein HOXA5; Homeobox protein M2; hox-1.3
Gene Aliases: Hox-1.3; HOX1; HOX1.3; HOX1C; Hoxa-5; HOXA5
UniProt ID: (Human) P20719, (Bovine) Q2HJ67, (Mouse) P09021
Entrez Gene ID: (Horse) 100054486, (Human) 3202, (Dog) 482370, (Bovine) 768039, (Rat) 79241, (Guinea pig) 100722624, (Mouse) 15402
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.