OriGene
Manufacturer Code:TA811550
Catalog # 50-168-4112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 40-276 of human HOXA3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: homeo box 1E; homeo box A3; Homeobox protein Hox-1E; Homeobox protein Hox-A3; Hox-1.5-like protein; HOX1 HOX1E
Gene Aliases: HOX1; HOX1E; HOXA3
UniProt ID: (Human) O43365
Entrez Gene ID: (Human) 3200
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.