Invitrogen
Manufacturer Code:PA579388
Catalog # PIPA579388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human HOXA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 1E; homeo box A3; homeobox A3; Homeobox protein Hox-1.5; Homeobox protein Hox-1E; Homeobox protein Hox-A3; Homeobox protein Hox-A3 Homeobox protein Hox-1E HOXA3 HOX1E; Homeobox protein MO-10; Hox-1.5-like protein
Gene Aliases: Hox-1.5; HOX1; HOX1E; Hoxa-3; HOXA3; Mo-10
UniProt ID: (Human) O43365, (Mouse) P02831
Entrez Gene ID: (Human) 3200, (Mouse) 15400, (Rat) 103690130
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.