Invitrogen
Manufacturer Code:PA568810
Catalog # PIPA568810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat HOXA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 1E; homeo box A3; homeobox A2; homeobox A3; Homeobox protein Hox-1.5; Homeobox protein Hox-1E; Homeobox protein Hox-A3; Homeobox protein MO-10; Hox-1.5-like protein; HOX1 HOX1E Hox-1.5-like protein homeo box 1E homeo box A3 homeobox protein Hox-1E homeobox protein Hox-A3
Gene Aliases: Hox-1.5; HOX1; HOX1E; Hoxa-3; HOXA3; Mo-10
UniProt ID: (Bovine) Q08DG7, (Human) O43365, (Mouse) P02831
Entrez Gene ID: (Rabbit) 100328784, (Bovine) 509991, (Pig) 100519809, (Human) 3200, (Dog) 482368, (Horse) 100054344, (Mouse) 15400, (Guinea pig) 100719597, (Rat) 103690130
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.