Invitrogen
Manufacturer Code:PA5101637
Catalog # PIPA5101637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HOXA11/D11 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E230017H14Rik homeo box 1I homeo box 4F homeo box A11 homeo box D11 homeobox A11 homeobox D11 homeobox protein Hox-1.9 Homeobox protein Hox-1I homeobox protein Hox-4.6 Homeobox protein Hox-4F homeobox protein Hox-5.5 homeobox protein HOXA11 Homeobox protein Hox-A11 homeobox protein Hox-D11 HOX1 Hox-1.9 HOX11 HOX1I HOX4 Hox-4.6 Hox-4.6 mouse homolog of HOX4F Hox-5.4 Hox-5.5 Hoxa11 Hoxa-11 HOXD11 Hoxd-11 LOC102553842 NEWGENE_1564605 RUSAT1 uncharacterized LOC102553842; homeo box 1I; homeo box 4F; homeo box A11; homeo box D11; Homeobox protein Hox-1.9; Homeobox protein Hox-1I; Homeobox protein Hox-4.6; Homeobox protein Hox-4F; Homeobox protein Hox-5.5; Homeobox protein Hox-A11; Homeobox protein Hox-D11; homeobox protein HOXA11; Hox-4.6, mouse, homolog of
Gene Aliases: E230017H14Rik; Hox-1.9; Hox-4.6; Hox-5.4; Hox-5.5; HOX1; HOX1I; HOX4; HOX4F; Hoxa-11; HOXA11; Hoxd-11; HOXD11; RUSAT1
UniProt ID: (Human) P31270, (Human) P31277, (Mouse) P31311, (Mouse) P23813
Entrez Gene ID: (Human) 3207, (Human) 3237, (Mouse) 15396, (Mouse) 15431
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.