Proteintech
Manufacturer Code:554951AP150UL
Catalog # 50-173-6184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeo box 1I; homeo box A11; homeobox A11 Homeobox protein Hox 1I Homeobox protein Hox A11 HOX1 HOX1I HOXA11; Homeobox protein Hox-1.9; Homeobox protein Hox-1I; Homeobox protein Hox-A11; homeobox protein HOXA11
Gene Aliases: Hox-1.9; HOX1; HOX1I; Hoxa-11; HOXA11; RUSAT1
UniProt ID: (Human) P31270, (Mouse) P31311
Entrez Gene ID: (Human) 3207, (Mouse) 15396
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.