Invitrogen
Manufacturer Code:PA538578
Catalog # PIPA538578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human HOXA1/B1/D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: early retinoic acid 1; homeo box 4G; homeo box A1; homeo box B1; homeo box D1; homeobox 1F; homeobox protein Hox-1.6; Homeobox protein Hox-1F; Homeobox protein Hox-2.9; Homeobox protein Hox-2I; Homeobox protein Hox-4.9; Homeobox protein Hox-A1; Homeobox protein Hox-B1; Homeobox protein Hox-D1; Homeobox protein Hox-GG; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; HOXA1 homeobox A1 BSAS HOX1 HOX1F MGC45232 HOX A1 homeodomain protein Hox 1.6-like protein homeo box A1 homeobox 1F homeobox protein HOX-A1 lab-like protein Homeobox protein Hox-B1 Homeobox protein Hox-2I HOX2I; lab-like protein
Gene Aliases: BSAS; ERA1; HCFP3; Hox-1.6; Hox-2.9; Hox-4.7; Hox-4.9; HOX1; HOX1F; HOX2; HOX2I; HOX4; HOX4G; HOXA1; Hoxb-1; HOXB1; Hoxd-1; HOXD1
UniProt ID: (Human) P49639, (Human) P14653, (Human) Q9GZZ0, (Mouse) P17919, (Mouse) Q01822
Entrez Gene ID: (Human) 3198, (Human) 3211, (Human) 3231, (Mouse) 15394, (Mouse) 15407, (Mouse) 15429
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.