Invitrogen
Manufacturer Code:PA568809
Catalog # PIPA568809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the c terminal region of mouse HOXA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: early retinoic acid 1; homeo box A1; homeobox 1F; homeobox A1; homeobox protein Hox-1.6; Homeobox protein Hox-1F; Homeobox protein Hox-A1; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; HOXA1 homeobox A1 BSAS HOX1 HOX1F MGC45232 HOX A1 homeodomain protein Hox 1.6-like protein homeo box A1 homeobox 1F homeobox protein HOX-A1 lab-like protein; lab-like protein
Gene Aliases: BSAS; ERA1; Hox-1.6; HOX1; HOX1F; HOXA1
UniProt ID: (Human) P49639
Entrez Gene ID: (Rabbit) 100328782, (Horse) 100068884, (Bovine) 537889, (Human) 3198, (Dog) 482367, (Guinea pig) 100717922, (Mouse) 15394, (Rat) 103690132
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.