Invitrogen
Manufacturer Code:PA536164
Catalog # PIPA536164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 180-220 of human Hox-A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: early retinoic acid 1; homeo box A1; homeobox 1F; homeobox A1; homeobox protein Hox-1.6; Homeobox protein Hox-1F; Homeobox protein Hox-A1; Homeobox protein Hox-A1 Homeobox protein Hox-1F HOXA1 HOX1F; homeoboxless protein ERA-1-399; homeotic protein ERA-1-993; Hox 1.6-like protein; HOX A1 homeodomain protein; lab-like protein
Gene Aliases: BSAS; ERA1; Hox-1.6; HOX1; HOX1F; HOXA1
UniProt ID: (Human) P49639
Entrez Gene ID: (Human) 3198, (Mouse) 15394, (Rat) 103690132
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.